missing translation for 'onlineSavingsMsg'
Learn More

LDB2 Antibody (1C3), Novus Biologicals™

Product Code. 18369308 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369308 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369308 Supplier Novus Biologicals Supplier No. H00009079M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LDB2 Monoclonal antibody specifically detects LDB2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDB2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1C3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH34019
Gene Alias Carboxyl-terminal LIM domain-binding protein 1, CLIM-1, CLIM1LDB1, LDB-2, LIM binding domain 2, LIM domain binding 2, LIM domain-binding factor CLIM1, LIM domain-binding factor-2, LIM domain-binding protein 2
Host Species Mouse
Immunogen LDB2 (AAH34019, 1 a.a. ~ 373 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSSTPHDPFYSSPFGPFYRRHTPYMVQPEYRIYEMNKRLQSRTEDSDNLWWDAFATEFFEDDATLTLSFCLEDGPKRYTIGRTLIPRYFSTVFEGGVTDLYYILKHSKESYHNSSITVDCDQCTMVTQHGKPMFTKVCTEGRLILEFTFDDLMRIKTWHFTIRQYRELVPRSILAMHAQDPQVLDQLSKNITRMGLTNFTLNYLRLCVILEPMQELMSRHKTYNLSPRDCLKTCLFQKWQRMVAPPAEPTRQPTTKRRKRKNSTSSTSNSSAGNNANSTGSKKKTTAANLSLSSQVPDVMVVGEPTLMGGEFGDEDERLITRLENTQYDAANGMDDEEDFNNSPALGNNSPWNSKPPATQETKSENPPPQASQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9079
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.