Learn More
Motic™ LD Plan Achromatic Objectives
Product Code. 17178598
Change view
Click to view available options
Magnification Power:
10X
20X
5X
Numerical Aperture:
0.13
0.25
0.4
Working Distance:
8.1 mm
17.5 mm
20.3 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Power Numerical Aperture Working Distance unitSize
17178598 5X 0.13 20.3 mm Each
17188598 10X 0.25 17.5 mm Each
17198598 20X 0.4 8.1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17178598 Supplier Motic™ Supplier No. 1101000203611
Only null left
Add to basket
Add to basket
Plan Achromatic objectives for use with TEC POL Microscopes.
  • For use with Motic™ Panthera TEC POL Microscopes
  • LD Plan Achromatic objectives
Specifications
Type Objective
For Use With (Equipment) Motic™ TEC POL Microscopes
Lens Type Plan Achromatic
Magnification Power 5X
Numerical Aperture 0.13
Working Distance 20.3 mm