missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCP2, Rabbit anti-Human, SureLight 488, Clone: 3F8, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB19085.100Reactions
This item is not returnable.
View return policy
Specifications
| LCP2 | |
| Monoclonal | |
| SureLight 488 | |
| In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
| SLP-76/SLP76 | |
| Rabbit | |
| 100 Reactions | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| 3F8 | |
| Flow Cytometry (5 uL/106 cells or 0.05 ug/mL) The optimal working dilution should be determined by the end user. | |
| lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of 76kDa) | |
| LCP2 | |
| A synthetic phosphopeptide corresponding to residues surrounding Y128 of human LCP2. | |
| RUO | |
| 3937 | |
| Store at 2-8°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction