missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCORL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555980
This item is not returnable.
View return policy
Description
Immunogen sequence: TYLHQLTLQK MVTQFKEKNE SLQYETSNPT VQLKIPQLRV SSVSKSQPDG SGLLDVMYQV SKTSSVLEGS ALQKLKNILP KQNKIECSGP VTHSSV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 27%.
May act as transcription activator that binds DNA elements with the sequence 5'-CCCTATCGATCGATCTCTACCT-3'. May play a role in spermatogenesis.
Specifications
| LCORL | |
| Polyclonal | |
| Unconjugated | |
| LCORL | |
| fi43c07; im:7138788; Lcorl; LCoR-like protein; LCORLp1; LCORLp2; ligand dependent nuclear receptor corepressor like; ligand dependent nuclear receptor corepressor-like; ligand-dependent nuclear receptor corepressor-like protein; MBLK1-related protein; mblk1-related protein 1; Mblk1-related protein-1; MLR1; RGD1561241; si:dkeyp-26a9.6; Transcription factor MLR1; wu:fi43c07 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 254251 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N3X6 | |
| LCORL | |
| Recombinant protein corresponding to Human LCORL. Recombinant protein control fragment (Product #RP-95540). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction