missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCOR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598797
This item is not returnable.
View return policy
Description
LCOR Polyclonal Antibody for Western Blot, ELISA
LCOR is a transcriptional corepressor widely expressed in fetal and adult tissues that is recruited to agonist-bound nuclear receptors through a single LxxLL motif, also referred to as a nuclear receptor (NR) box.
Specifications
| LCOR | |
| Polyclonal | |
| Unconjugated | |
| LCOR | |
| 3110023F06Rik; A630025C20Rik; C10orf12; KIAA1795; Lcor; ligand dependent nuclear receptor corepressor; ligand-dependent corepressor; Mblk1-related protein 2; Mblk1-related protein-2; mKIAA1795; MLR2; RGD1311704; transcription factor MLR2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 84458 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4.57 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q96JN0 | |
| LCOR | |
| Recombinant Human Ligand-dependent corepressor protein (13-144AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction