missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCN6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102384
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LCN6.
LCN6 is a member of the lipocalin family of hydrophobic ligand binding proteins and exhibit conserved beta strand basket like structures. LCN6 may play a role in male fertility.
Specifications
| LCN6 | |
| Polyclonal | |
| Unconjugated | |
| LCN6 | |
| 9230101D24Rik; epididymal-specific lipocalin ELP16; epididymal-specific lipocalin LCN6; epididymal-specific lipocalin-6; hLcn5; LCN5; Lcn6; lipocalin 5; lipocalin 6; lipocalin-5; MCG18768; UNQ643; UNQ643/PRO1273 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 158062, 414138, 620709 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62502 | |
| LCN6 | |
| A synthesized peptide derived from human LCN6(Accession P62502), corresponding to amino acid residues A48-E98. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction