missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LCMT1 Protein

Product Code. 18488880 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
100μg
Unit Size:
0.50mg
100µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18488880 0.5 mg 0.50mg
18323731 100μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18488880 Supplier Novus Biologicals™ Supplier No. NBP1788450.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-334 of Human LCMT1 The Recombinant Human LCMT1 Protein is derived from E. coli. The Recombinant Human LCMT1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20 mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 1 mM DTT, 0.15 M NaCl.
Molecular Weight (g/mol) 41 kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
Source Recombinant human LCMT7 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen LCMT1, 1-334 aa, Human, His tag E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name LCMT1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.