missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Chemical Name or Material | LCMS Daily Check Standard 1 |
| Name Note | EPA 535 |
| Packaging | Glass Ampoule |
| CAS | 58-08-2, 50-55-5 |
| Explosion Limit | 4.4% (volume); 16% (volume) |
| Ignition Point | 525°C |
| Vapor Pressure | 97 hPA (20°C) |
| Analyte or Component Names | Caffeine,Reserpine |
| Density | 0.7822g/cm3 |
| Physical Form | Fluid |
| Show More |
Safety and Handling
Product Identifier
- LCMS Daily Check
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: acetonitrile
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?