missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCLAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561911
This item is not returnable.
View return policy
Description
Immunogen sequence: ICLKASLKGV PGFGWAMQAA AYIFIHRKWK DDKSHFEDMI DYFCDIHEPL QLLIFPEGTD LTENSKSRSN AFAEKNGLQK YEYV Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
Exhibits acyl-CoA:lysocardiolipin acyltransferase (ALCAT) activity; catalyzes the reacylation of lyso-cardiolipin to cardiolipin (CL), a key step in CL remodeling. Recognizes both monolysocardiolipin and dilysocardiolipin as substrates with a preference for linoleoyl-CoA and oleoyl-CoA as acyl donors. Also exhibits 1-acyl-sn-glycerol-3-phosphate acyltransferase activity (AGPAT) activity; converts 1-acyl-sn-glycerol-3- phosphate (lysophosphatidic acid or LPA) into 1,2-diacyl-sn-glycerol-3- phosphate (phosphatidic acid or PA) by incorporating an acyl moiety at the sn-2 position of the glycerol backbone (PubMed:16620771). Possesses both lysophosphatidylinositol acyltransferase (LPIAT) and lysophosphatidylglycerol acyltransferase (LPGAT) activities (PubMed:19075029). Required for establishment of the hematopoietic and endothelial lineages. [UniProt].
Specifications
| LCLAT1 | |
| Polyclonal | |
| Unconjugated | |
| LCLAT1 | |
| 1-acylglycerol-3-phosphate O-acyltransferase 8; 1-AGP acyltransferase 8; 1-AGPAT 8; 1AGPAT8; Acyl-CoA:lysocardiolipin acyltransferase 1; Agpat8; AI181996; Alcat1; Gm91; HSRG1849; LCLAT1; Lycat; lysocardiolipin acyltransferase 1; RGD1565906; UNQ1849; UNQ1849/PRO3579 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 253558 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| Lot-Specific | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UWP7 | |
| LCLAT1 | |
| Recombinant protein corresponding to Human LCLAT1. Recombinant protein control fragment (Product #RP-95707). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction