missing translation for 'onlineSavingsMsg'
Learn More

LCL161, MedChemExpress

Product Code. 16429167
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429167 5 mg 5mg
16426598 10 mg 10mg
16452687 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16429167 Supplier Medchem Express Supplier No. HY15518

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LCL161 is a IAP inhibitor which inhibits XIAP in HEK293 cell and cIAP1 in MDA-MB-231 cell with IC50s of 35 and 0.4 nM, respectively.

LCL161 is a IAP inhibitor which inhibits XIAP in HEK293 cell and cIAP1 in MDA-MB-231 cell with IC50s of 35 and 0.4 nM, respectively.

Specifications

Chemical Name or Material LCL161
CAS 1005342-46-0
Color White
Purity Grade Notes Research
Molecular Formula C26H33FN4O3S
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (199.75 mM; Need ultrasonic)
SMILES C[C@H](NC)C(N[C@@H](C1CCCCC1)C(N2[C@H](C3=NC(C(C4=CC=C(F)C=C4)=O)=CS3)CCC2)=O)=O
Molecular Weight (g/mol) 500.63
Formula Weight 500.63
Percent Purity 98.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.