missing translation for 'onlineSavingsMsg'
Learn More

Lck Inhibitor, MedChemExpress

Product Code. 16477707
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477707 5 mg 5mg
16445746 100 mg 100mg
16434978 10 mg 10mg
16401227 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477707 Supplier Medchem Express Supplier No. HY12072
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lck Inhibitor is a potent, orally active Lck (lymphocyte specific kinase) inhibitor with IC50s of 7, 2.1, 4.2 and 200 nM for Lck, Lyn, Src and Syk kinases, respectively. Lck Inhibitor shows >1000-fold selectivity for Lck over MAPK, CDK and RSK family representatives. Lck Inhibitor inhibits T cell proliferation and in vivo models of arthritis.

Lck Inhibitor is a potent, orally active Lck (lymphocyte specific kinase) inhibitor with IC50s of 7, 2.1, 4.2 and 200 nM for Lck, Lyn, Src and Syk kinases, respectively. Lck Inhibitor shows >1000-fold selectivity for Lck over MAPK, CDK and RSK family representatives. Lck Inhibitor inhibits T cell proliferation and in vivo models of arthritis.

Specifications

Chemical Name or Material Lck Inhibitor
CAS 847950-09-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H30N8O
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (188.46 mM; Need ultrasonic)
SMILES O=C1N(C2=C(C)C=CC=C2C)C3=NC4=CC=CC=C4N3C5=NC(NC6=CC=C(N7CCN(C)CC7)C=C6)=NC=C15
Molecular Weight (g/mol) 530.62
Formula Weight 530.62
Percent Purity 98.2%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.