missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ LCK Cuvette Tests

Product Code. 10266031
Click to view available options
Range:
1 to 70, 70 to 1000 mg/L Cl
Unit Size:
Pack of 24
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10266031

Brand: Hach Lange™ LCK311

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Pre-dosed reagents for maximum safety. Easy to use and photometric evaluation, packaged in a fully equipped box. Environmentally friendly due to less chemicals.

  • Chloride cuvette test 1-70 mg/L / 70-1000 mg/L Cl
  • For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
  • Shelf life of 12 months from production date
  • 2 measuring ranges with 1 test
  • Precise and reliable measurement values
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of color-coding

Specifications

For Use With (Application) Chloride Cuvette Test
For Use With (Equipment) Photometric Tests
Parameter Specifications Chloride
Range 1 to 70, 70 to 1000 mg/L Cl
Test Method Iron(III)-Thiocyanate
Type LCK Cuvette Test
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • LCK 311 Chlorid/Chloride/Chlorure, Sample cuvette
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after single exposure Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H370-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : methanol, Nitric acid ... %
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.