missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lck Antibody (33D196), Janelia Fluor™ 669, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-56678JF669
This item is not returnable.
View return policy
Description
Lck Monoclonal antibody specifically detects Lck in Human,Mouse samples. It is validated for Western Blot
Specifications
| Lck | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EC 2.7.10, EC 2.7.10.2, Leukocyte C-terminal Src kinase, LSK, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16 | |
| This monoclonal antibody was raised against a synthetic peptide containing amino acid residues of human lck protein. | |
| 0.2 mL | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 3932 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| 33D196 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction