missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCHN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566393
This item is not returnable.
View return policy
Description
Immunogen sequence: FYVNVADIESL EVEVSYVACT TEKIFEEKRE LYDVYVDNQN VKTHHDHLQP LLKINSADRE KYRRLNEQRQ MLL Highest antigen sequence identity to the following orthologs - mouse 100%, rat 99%.
Probable guanine nucleotide exchange factor (GEF). May promote the exchange of GDP to GTP, converting inactive GDP-bound small GTPases into their active GTP-bound form (Probable). May play a role in neuritogenesis, as well as in neuronal recovery and/or restructuring in the hippocampus following transient cerebral ischemia. [UniProt].
Specifications
| LCHN | |
| Polyclonal | |
| Unconjugated | |
| DENND11 | |
| AI841796; DENN domain containing 11; DENN domain-containing protein 11; Dennd11; E330009J07Rik; KIAA1147; LCHN; mKIAA1147; predicted protein of HQ2561; PRO2561; protein LCHN; RGD1563986; RIKEN cDNA E330009J07 gene; si:ch211-238n5.4; similar to RIKEN cDNA E330009J07 gene; wu:fa55h08 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57189 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A4D1U4 | |
| DENND11 | |
| Recombinant Human LCHN. Recombinant protein control fragment (Product #RP-107066). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction