missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCAT Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00108AF488
This item is not returnable.
View return policy
Description
LCAT Polyclonal antibody specifically detects LCAT in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| LCAT | |
| Polyclonal | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 2.3.1.43, lecithin-cholesterol acyltransferasePhospholipid-cholesterol acyltransferase, phosphatidylcholine-sterol acyltransferase | |
| Produced in rabbits immunized with purified, recombinant Human LCAT (NP_000220.1; Met1-Glu440). | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 3931 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction