Learn More
Olympus™ LCACHN 40XIPC 40X Achromat Dry Pre-Centered Phase Objective
Product Code. 15753258
Change view
Click to view available options
Magnification Power:
40X
Unit Size:
Each
Product Code. Magnification Power unitSize
15753258 40X Each
1 options
Product Code. 15753258 Supplier Olympus™ Supplier No. N5229400
  • ONLINE EXCLUSIVE PRICE
    £682.50
    £607.00 / Pack of 1
    Save £75.50
    11% Off
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
Corrected for chromatic and spherical aberration.
  • Corrected for axial chromatic aberration.
  • Corrected for spherical aberration.
  • Requires CKX3-SLP pre-centered slider.
Specifications
Type Achromat Dry Pre-Centered Phase
Connector Types RMS Threading
Description Field number: 22
Focus Distance Working distance: 2.2 mm
Magnification Power 40X
Numerical Aperture 0.55