Learn More
Olympus™ LCACHN 20XIPC 20X Achromat Dry Pre-Centered Phase Objective
Click to view available options
Magnification Power:
20X
Unit Size:
Each
Description
- Corrected for axial chromatic aberration.
- Corrected for spherical aberration.
- Requires CKX3-SLP pre-centered slider.
Specifications
Specifications
| Type | Achromat Dry Pre-Centered Phase |
| Connector Types | RMS Threading |
| Description | Field number: 22 |
| Focus Distance | Working distance: 3.2 mm |
| Magnification Power | 20X |
| Numerical Aperture | 0.4 |