missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC8/DYNLL1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18130-1-AP-150UL
This item is not returnable.
View return policy
Description
The LC8/DYNLL1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LC8/DYNLL1. This antibody recognizes human, mouse, rat antigen. The LC8/DYNLL1 antibody has been validated for the following applications: WB, IHC, IF/ICC, FC (Intra), IP, ELISA analysis.Specifications
| LC8/DYNLL1 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8655 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), Immunoprecipitation, ELISA | |
| 450 μg/mL | |
| DLC1; DLC8; DNCL1; DNCLC1; LC8; LC8a; MGC126137; MGC126138; PIN; hdlc1 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 8-10 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction