Learn More
LC3C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18726-1-AP-150UL
 View more versions of this product
Product Code. 16868604
  • £349.00 / 150µL
Description
The antibody is specific to the propeptide of LC3C.
MAP1LC3C, also named as LC3C, belongs to the MAP1 LC3 family. It is the human homolog of yeast Atg8 and is involved in the formation of autophagosomal vacuoles, called autophagosomes. LC3C is produced by the proteolytic cleavage after the conserved C-terminal Gly residue. The antibody is specific to the prepeptide of LC3C.
Specifications
Specifications
LC3C
Polyclonal
Unconjugated
MAP1LC3C
LC3C, MAP1A/MAP1B LC3 C, MAP1A/MAP1B light chain 3 C, MAP1LC3C
Rabbit
Antigen Affinity Chromatography
RUO
440738
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BXW4
MAP1LC3C
Peptide
150 μL