missing translation for 'onlineSavingsMsg'
Learn More

LC3C Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18374574 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374574 20 μL 20µL
18320987 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18374574 Supplier Novus Biologicals Supplier No. NBP30328020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LC3C Polyclonal specifically detects LC3C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LC3C
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ATG8J, Autophagy-related protein LC3 C, Autophagy-related ubiquitin-like modifier LC3 C, LC3C, LC3-like protein 2, MAP1 light chain 3-like protein 2, MAP1 light chain 3-like protein 3, MAP1A/MAP1B LC3 C, microtubule-associated protein 1 light chain 3 gammaMAP1A/MAP1B light chain 3 C, microtubule-associated proteins 1A/1B light chain 3C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human LC3C (NP_001004343.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Autophagy, Cellular Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 440738
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.