missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LC3B Polyclonal Antibody, DyLight™ 550

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522940
This item is not returnable.
View return policy
Description
LC3B Polyclonal Antibody for Flow, ICC/IF, IHC (F), IHC (Free), IHC (P), IM, IP, Western Blot
LC3B (Autophagy Marker Light Chain 3B, MAP1A/MAP1B LC3 B) in humans, is encoded by the gene MAP1LC3B (Microtubule-associated proteins 1A/1B light chain 3B). LC3B is associated with microtubule assembly and important is in neurogenesis. Recent studies indicate that LC3B plays a vital role in autophagy, a process that involves the bulk degradation of cytoplasmic component. Three human LC3 isoforms undergo post-translational modifications during autophagy. Macroautophagy is the major inducible pathway for the general turnover of cytoplasmic constituents in eukaryotic cells, it is also responsible for the degradation of active cytoplasmic enzymes and organelles during nutrient starvation. Macroautophagy involves the formation of double-membrane bound autophagosomes which enclose the cytoplasmic constituent targeted for degradation in a membrane bound structure, which then fuse with the lysosome (or vacuole) releasing a single-membrane bound autophagic bodies which are then degraded within the lysosome (or vacuole). LC3B is a microtubule-associated protein that mediate the physical interactions between microtubules and components of the cytoskeleton. LC3B may play a role in processes involving cancer, aging, metabolic and neurodegenerative disorders and cardiovascular/pulmonary diseases.
Specifications
| LC3B | |
| Polyclonal | |
| DyLight 550 | |
| MAP1LC3B2 | |
| 1010001C15Rik; Atg8; ATG8E; ATG8F; ATG8G; Autophagy-related protein LC3 B; autophagy-related ubiquitin-like modifier LC3 B; LC3; LC3 beta; LC3A; LC3B; light chain 3; MAP1 light chain 3-like protein 2; MAP1A/1B light chain 3 B; MAP1A/1BLC3; MAP1A/MAP1B LC3 A; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 B; Map1alc3; MAP1BLC3; MAP1LC3; MAP1LC3A; MAP1LC3B; MAP1LC3B2; MAP1LC3B-a; microtubule associated protein 1 light chain 3 beta; microtubule associated protein 1 light chain 3 beta 2; microtubule-associated protein 1 light chain 3; microtubule-associated protein 1 light chain 3 beta; microtubule-associated protein 1 light chain 3 beta 2; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3 beta 2; Microtubule-associated proteins 1A/1B light chain 3B; Microtubule-associated proteins 1A/1B light chain 3B-like; Microtubule-associated proteins 1A/1B light chain 3B-like protein; Mpl3; RP11-346K17.1; wu:fb60g11; zbs559; zgc:56434 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100359928, 100462754, 322425, 408001, 479619, 643246, 64862, 67443, 81631 | |
| 4°C, store in dark | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.1 mg/mL | |
| 50mM sodium borate with 0.05% sodium azide | |
| A6NCE7, O41515, Q62625, Q9CQV6, Q9GZQ8 | |
| MAP1LC3B, MAP1LC3B2 | |
| Synthetic peptide made to the N-terminal region of the human LC3B protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Bovine, Pig, Yeast, Bacteria, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction