missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3B Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-1384FR
This item is not returnable.
View return policy
Description
LC3B Polyclonal antibody specifically detects LC3B in Human, Mouse, Rat, Porcine, Bacteria, Bovine, Canine, Primate, Yeast, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Electron Microscopy, Immunohistochemistry Free-Floating, Gene Knock-Out, KnockDown
Specifications
| LC3B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bacteria | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DyLight 680 | |
| ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
| Polyclonal LC3B Antibody was made to a synthetic peptide made to the N-terminal region of the human LC3B protein. [Uniprot: Q9GZQ8] | |
| 0.1 mL | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Neuroscience | |
| 81631 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction