missing translation for 'onlineSavingsMsg'
Learn More

LC3B Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1218 publications

£248.00 - £516.00

Specifications

Antigen LC3B
Concentration 1.0 mg/mL
Dilution Western Blot 0.5 - 2.0 ug/mL, Simple Western 1:50, Flow Cytometry, ELISA, Immunohistochemistry 1:200 - 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 20 ug/500 ug of protein, Immunohistochemistry-Paraffin 1:200 - 1:400, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay, SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
Applications ChIP Assay, ELISA, Flow Cytometry, Immunoassay, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In situ PLA, KnockDown
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18777963
View Documents
Novus Biologicals
NB100-2220SS
0.025 mL
£248.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18727253
View Documents
Novus Biologicals
NB100-2220
0.1 mL
£516.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LC3B Polyclonal specifically detects LC3B in Human, Mouse, Rat, Porcine, Alligator, Avian, Bacteria, Bovine, Canine, Chicken, Chinese Hamster, Guinea Pig, Hamster, Invertebrate, Monkey, Primate, Rabbit, Golden Syrian Hamster, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay, SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Knockdown Validated.
Specifications

Specifications

LC3B
Western Blot 0.5 - 2.0 ug/mL, Simple Western 1:50, Flow Cytometry, ELISA, Immunohistochemistry 1:200 - 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 20 ug/500 ug of protein, Immunohistochemistry-Paraffin 1:200 - 1:400, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay, SDS-Page, Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated
Polyclonal
Rabbit
Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Mitophagy, Neuroscience
PBS with 0.02% Sodium Azide
anti-LC3B, ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, LC3B ihc, LC3B immunohistochemistry, LC3B immunoprecipitation, LC3B western blot, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B
MAP1LC3B
IgG
Affinity Purified
Human, rat, mouse, zebrafish, bacteria, hamster, and porcine. Bovine reactivity reported in scientific literature (PMID: 24895572) Primate reactivity reported in scientific literature (PMID: 25142602) The mouse detection has been reported to be weaker than the human. Immunogen sequence has 84% homology to Xenopus.
1.0 mg/mL
ChIP Assay, ELISA, Flow Cytometry, Immunoassay, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In situ PLA, KnockDown
Unconjugated
RUO
Human, Mouse, Rat, Pig, Alligator, Avian, Bacteria, Bovine, Canine, Chicken, Guinea Pig, Hamster, Invertebrate, Monkey, Primate, Rabbit, Golden Syrian Hamster, Zebrafish
Q9GZQ8
81631
Polyclonal LC3B Antibody was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [UniProt# Q9GZQ8]
Primary
Store at -20°C.
14.688 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.