missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3B Antibody (7D11), Novus Biologicals™
Rabbit Monoclonal Antibody
£269.00 - £431.00
Specifications
| Antigen | LC3B |
|---|---|
| Clone | 7D11 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30026743
|
Novus Biologicals
NBP3-26154-50ul |
50 μL |
£269.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028779
|
Novus Biologicals
NBP3-26154-100ul |
100 μL |
£431.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LC3B Monoclonal antibody specifically detects LC3B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
| LC3B | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
| A synthesized peptide derived from Human LC3B [UniProt Q9GZQ8] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 7D11 | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 81631 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title