missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3B Antibody (1251D) - BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-59800SS
This item is not returnable.
View return policy
Description
LC3B Monoclonal specifically detects LC3B in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockout Validated.
Specifications
| LC3B | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 - 2 ug/ml, Simple Western 5 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Flow (Intracellular) 1 ug/ml, Knockout Validated | |
| anti-LC3B, ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, LC3B ihc, LC3B immunohistochemistry, LC3B immunoprecipitation, LC3B monoclonal, LC3B western blot, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
| Rabbit | |
| 14.688 kDa | |
| 0.025 mg | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Mitophagy, Neuroscience | |
| 81631 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry, KnockDown | |
| 1251D | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| MAP1LC3B | |
| Recombinant monoclonal LC3B Antibody (1251D) was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [Uniprot: Q9GZQ8]. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction