missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3B Antibody (1251A) - BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody has been used in 9 publications
Brand: Novus Biologicals NBP2-46892
This item is not returnable.
View return policy
Description
LC3B Monoclonal specifically detects LC3B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), Knockout Validated.
Specifications
| LC3B | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunoprecipitation 2-10 ug, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen reported in scientific literature (PMID 32278100), Flow (Intracellular) 1 - 2.5 ug/mL, Knockout Validated | |
| Q9GZQ8 | |
| MAP1LC3B | |
| Recombinant monoclonal LC3B Antibody (1251A) was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [UniProt# Q9GZQ8]. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Specific for LC3B. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay | |
| 1251A | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, LC3B monoclonal, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
| Rabbit | |
| 14.688 kDa | |
| 0.1 mg | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Mitophagy, Neuroscience | |
| 81631 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction