missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18722-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to LC3A.It recognize both LC3A-I and LC3A-II.
MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| LC3A | |
| Polyclonal | |
| Unconjugated | |
| MAP1LC3A | |
| LC3, LC3A, MAP1A/MAP1B light chain 3 A, MAP1ALC3, MAP1BLC3, MAP1LC3A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362245, 66734, 84557 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6XVN8, Q91VR7, Q9H492 | |
| Map1lc3a | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction