missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3A/LC3B Polyclonal Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522729
This item is not returnable.
View return policy
Description
The target sequence has 100% sequence homology with xenopus, bovine and zebrafish.
Autophagy is a process of intracellular bulk degradation in which cytoplasmic components, including organelles, are sequestered within double-membrane vesicles that deliver the contents to the lysosome/vacuole for degradation. During macroautophagy, the sequestering vesicles, termed autophagosomes, fuse with the lysosome or vacuole resulting in the delivery of an inner vesicle (autophagic body) into the lumen of the degradative compartment. There are 16 proteins participating in the autophagy pathway in human. The autophagy protein LC3, a mammalian homologue of Atg8, was originally identified as microtubule-associated protein 1 light chain 3. It is a component of both the MAP1A and MAP1B microtubule-binding domains and the heavy-chain independent regulation of LC3 expression might modify MAP1 microtubule-binding activity during development. LC3 is the only known mammalian protein identified that stably associates with the autophagosome membranes. LC3-I is cytosolic and LC3-II is membrane bound and enriched in the autophagic vacuole fraction. The detection of LC3-I to LC3-II conversion is a useful and sensitive marker for distinguishing autophagy in mammalian cells.
Specifications
| LC3A/LC3B | |
| Polyclonal | |
| Biotin | |
| MAP1LC3B2 | |
| 1010001C15Rik; 1010001H21Rik; 4922501H04Rik; Atg8; ATG8E; ATG8F; Autophagy-related protein LC3 A; autophagy-related protein LC3 B; Autophagy-related ubiquitin-like modifier LC3 A; autophagy-related ubiquitin-like modifier LC3 B; LC3; LC3a; LC3b; MAP1 light chain 3-like protein 1; MAP1 light chain 3-like protein 2; MAP1A/1B light chain 3 A; MAP1A/1B light chain 3 B; MAP1A/1BLC3; MAP1A/MAP1B LC3 A; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 A; MAP1A/MAP1B light chain 3 B; Map1alc3; MAP1BLC3; Map1lc3; Map1lc3a; MAP1LC3B; MAP1LC3B-a; microtubule associated protein 1 light chain 3 alpha; microtubule associated protein 1 light chain 3 beta; microtubule-associated protein 1 light chain 3; microtubule-associated protein 1 light chain 3 alpha; Microtubule-associated protein 1 light chain 3 beta; microtubule-associated protein 1-light chain 3A; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3A; Microtubule-associated proteins 1A/1B light chain 3B; Mpl3; wu:fb60g11; zbs559; zgc:56434; zgc:77094 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100359928, 322425, 362245, 406466, 477201, 479619, 643246, 64862, 66734, 67443, 81631, 84557 | |
| 4° C, store in dark | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 0.65 mg/mL | |
| PBS with 0.05% sodium azide | |
| A6NCE7, Q62625, Q6XVN8, Q91VR7, Q9CQV6, Q9GZQ8, Q9H492 | |
| MAP1LC3A, MAP1LC3B, MAP1LC3B2 | |
| A synthetic peptide made to an internal portion of the human LC3 protein sequence (between residues 25-121). | |
| 100 μL | |
| Primary | |
| Amphibian, Canine, Fish, Human, Mouse, Plant, Rat, Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction