missing translation for 'onlineSavingsMsg'
Learn More

LC3A Antibody (2312D) [Biotin], Novus Biologicals™

Product Code. 30737779 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737779 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737779 Supplier Novus Biologicals Supplier No. FAB9915B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

LC3A Monoclonal antibody specifically detects LC3A in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LC3A
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2312D
Conjugate Biotin
Formulation PBS
Gene Alias ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, MAP1 light chain 3-like protein 1, MAP1A/1B light chain 3 A, MAP1A/MAP1B LC3 A, MAP1A/MAP1B light chain 3 A, MAP1ALC3, MAP1BLC3, MAP1LC3A, Microtubule-associated protein 1 light chain 3 alpha, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A
Host Species Rabbit
Immunogen Synthetic peptide containing human LC3A peptide, Accession # Q9H492
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Macroautophagy, Membrane Trafficking and Chaperones, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84557
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.