missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3A Antibody (2312D) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-80828
This item is not returnable.
View return policy
Description
LC3A Monoclonal specifically detects LC3A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LC3A | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry : 3-25 μg/mL, Immunohistochemistry-Paraffin : 3-25 μg/mL | |
| ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2312D | |
| Unconjugated | |
| PBS | |
| MAP1LC3A | |
| Partial synthetic peptide from the C-terminal of human LC3A [UniProt Q9H492] | |
| 0.1 mg | |
| Autophagy, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Macroautophagy, Neuroscience, Phospho Specific, Signal Transduction | |
| 84557 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction