Learn More
Trajan Scientific and Medical™ LC PEEKsil™ Tubing
Mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems.
Marque: Trajan Scientific and Medical™ 0624290
Code : NEW
Informations supplémentaires : Weight : 0.01000kg
Description
- Fused silica internal wall provides smooth surface, free of imperfections
- PEEKsil compatible with most organic solvents and resistant to strong acids
- Effective pH range of 0 to 10
- Flexibility ensures easy connection between columns, detectors and injectors
Spécification
Tubing | |
0.075 mm | |
50 mm |
5/Pk. | |
1.6 mm | |
Polymer-sheathed Fused Silica |