missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LC 2
Description
LC 2 is a mutant-selective PROTAC™ Degrader of KRAS, comprising a ligand for the von Hippel Lindau E3 ligase joined to the KRAS inhibitor MRTX849. LC 2 induces selective degradation of KRASG12C in cancer cell lines without inducing degradation of any other mutants (DC50 = 0.25-0.76 μM), leading to suppression of MAPK signaling and modulation of ERK signaling in both homozygous and heterozygous KRASG12C cancer cell lines. LC 2 inhibits phosphorylation of ERK in SW1573 cell lines.
Negative control LC 2 Epimer (Cat. No. 7421) also available.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2 S,4 R)-1-(-2-(3-(3-(-2-(((7-(8-Chloronaphthalen-1-yl)-4-(-3-(cyanomethyl)-4-(2-fluoroacryloyl)piperazin-1-yl)-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidin-2-yl)oxy)methyl)pyrrolidin-1-yl)propoxy)propanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-m) |
| CAS | 2502156-03-6 |
| Quantity | 2 mg |
| Target | Active Degraders |
| Molecular Formula | C59H71ClFN11O7S |
| Purity | 0.95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?