missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LBX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55646-25ul
This item is not returnable.
View return policy
Description
LBX1 Polyclonal specifically detects LBX1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| LBX1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL | |
| homeobox, HPX6, HPX-6, ladybird homeobox 1, ladybird homeobox homolog 1, ladybird homeobox homolog 1 (Drosophila), Ladybird homeobox protein homolog 1, LBX1Hlady bird-like homeobox, transcription factor LBX1, transcription factor similar to D. melanogaster homeodomain protein lady birdlate | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10660 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| LBX1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ITWFQNRRAKLKRDLEEMKADVESAKKLGPSGQMDIVALAELEQNSEATAGGGGGCGRAKSRPGSPVLPPGAPKAPGAGALQLSPASPLTDQPASSQDCSEDEEDEEIDVD | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction