Learn More
LBR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12398-1-AP-20UL
 View more versions of this product
Product Code. 16810104
  • £135.00 / 20µL
Description
The protein encoded by this gene belongs to the ERG4/ERG24 family. It localized in the nuclear envelope inner membrane and anchors the lamina and the heterochromatin to the membrane. It may mediate interaction between chromatin and lamin B. Mutations of this gene has been associated with autosomal recessive HEM/Greenberg skeletal dysplasia. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.
Specifications
Specifications
LBR
Polyclonal
Unconjugated
LBR
DHCR14B, lamin B receptor, LBR, LMN2R, PHA
Rabbit
Antigen Affinity Chromatography
RUO
3930, 89789, 98386
-20°C
Liquid
Immunoprecipitation, Immunofluorescence, Flow Cytometry, Immunocytochemistry, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O08984, Q14739, Q3U9G9
LBR
LBR Fusion Protein Ag3088
20 μL
Primary
Rat, Mouse, Human