Learn More
LBR Mouse, anti-Human, Clone: 1A1B7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68378-1-IG-150UL
This item is not returnable.
View return policy
Description
The LBR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LBR. This antibody recognizes human, rat, mouse antigen. The LBR antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| LBR | |
| 1A1B7 | |
| DHCR14B, FLJ43126, lamin B receptor, LBR, LMN2R, PHA | |
| 150 μL | |
| 3930 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse | |
| 150 kDa | |
| IgG1 |