missing translation for 'onlineSavingsMsg'
Learn More

LBP Antibody [PerCP], Novus Biologicals™

Product Code. 30731331 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731331 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731331 Supplier Novus Biologicals Supplier No. AF6635C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

LBP Polyclonal antibody specifically detects LBP in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LBP
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias lipopolysaccharide binding protein, lipopolysaccharide-binding protein, LPS-binding protein, MGC22233
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse LBP, Gly25-Val481 (Ser102Arg, Tyr284His), Accession # NP_032515
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3929
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.