missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LBHD1 Polyclonal specifically detects LBHD1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | LBHD1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | C11orf48, chromosome 11 open reading frame 48, hypothetical protein LOC79081, MGC2477 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human LBHD1 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?