missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ LB Broth (Miller Luria-Bertani Broth)

Product Code. 12841670
Change view
Click to view available options
Format:
Pre-mixed 1 g capsules
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
12841670 Pre-mixed 1 g capsules Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12841670 Supplier Fisher BioReagents™ Supplier No. 12841670
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

LB Broth (Miller Luria-Bertani Broth) is supplied in containers of convenient pre-measured, pre-mixed 1 g capsules, and is for the growth and maintenance of Escherichia coli.

  • Pre-Buffered, 1 g capsules
  • pH 7.0
  • For growth and maintenance of E. coli

Specifications

Content And Storage Room temperature
Format Pre-mixed 1 g capsules
Preparation Method Use 25 capsules to make 1000 mL broth
For Use With (Application) Growth and Maintenance of E. Coli
pH 7
Solubility Soluble in boiling water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.