missing translation for 'onlineSavingsMsg'
Learn More

Melford LB Agar, Low Salt, Powder

Product Code. 30018400
Click to view available options
Quantity:
100 g
500 g
1 kg
2 kg
Unit Size:
100g
1kg
2kg
500g
This item is not returnable. View return policy

Product Code. 30018400

Brand: Melford L240302000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Molecular Biology Grade.

Molecular Biology Grade. For propagation and maintenance of E. Coli. Low Salt Formula.

Dissolve 35 grams in 1 Liter distilled water. Adjust the pH to 7.2 Sterilize by autoclaving for 15 minutes

Ingredients:

Casein Peptone: 10 g/L

Sodium Chloride: 5 g/L

Yeast Extract: 5 g/L

Agar: 15 g/L

Specifications

Quantity 2 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.