missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ LB-Agar, Capsules, Molecular Biology Certified™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
11 g
227 g
454 g
Format:
Capsule
Unit Size:
11g
227g
454g
Description
- Nutritionally rich medium originally developed for growth and maintenance of E. coli strains
- Eliminates the waste, inaccuracies, and mess associated with weighing out bulk powder
- Contains essential electrolytes for transport and osmotic balance, due to NaCl component
Specifications
Specifications
| Certifications/Compliance | Molecular Biology Certified™ FDA |
| Content And Storage | 15°C to 30°C |
| Format | Capsule |
| Grade | Molecular Biology |
| Product Type | Agar |
| Formulation | Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L |
| For Use With (Application) | For growth of bacteria |
| Quantity | 454 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction