missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lazabemide, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Lazabemide (Ro 19-6327) is a selective, reversible inhibitor of monoamine oxidase B (MAO-B) (IC50=0.03 μM) but less active for MAO-A (IC50>100 μM). Lazabemide inhibits monoamine uptake at high concentrations, the IC50 values are 86 μM, 123 μM and >500 μM for noradrenalin, serotonin and dopamine uptake, respectively. Lazabemide can be used for the research of parkinson and alzheimer′s disease.
Specifications
Specifications
| Chemical Name or Material | Lazabemide |
| CAS | 103878-84-8 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C8H10ClN3O |
| Quantity | 50 mg |
| Synonym | Ro 19-6327 |
| Solubility Information | DMSO : 5 mg/mL (25.05 mM; Need ultrasonic) |
| SMILES | ClC1=CC=C(C(NCCN)=O)N=C1 |
| Molecular Weight (g/mol) | 199.64 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction