missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Lazabemide hydrochloride
GREENER_CHOICE

Product Code. 18075032
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18075032 10 mg 10mg
18024282 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18075032 Supplier Tocris Bioscience™ Supplier No. 2460/10
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective MAO-B inhibitor

Selective, reversible monoamine oxidase B (MAO-B) inhibitor (IC50 values are 0.03 and > 100 μM for MAO-B and MAO-A respectively). Inhibits monoamine uptake at high concentrations (IC50 values are 86, 123 and > 500 μM for noradrenalin, serotonin and dopamine uptake respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103878-83-7
Molecular Formula C8H11Cl2N3O

Specifications

Quantity 10 mg
Formula Weight 236.1
Purity >98%
Molecular Weight (g/mol) 236.096
Chemical Name or Material Lazabemide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.