missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAYN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20535-1-AP-150UL
This item is not returnable.
View return policy
Description
Layilin is a receptor for hyaluronate.Specifications
| LAYN | |
| Polyclonal | |
| Unconjugated | |
| LAYN | |
| layilin, LAYN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 143903, 244864 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6UX15, Q8C351 | |
| LAYN | |
| LAYN Fusion Protein Ag14508 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction