missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Layilin Monoclonal Antibody (K94011_7A4), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | Layilin |
|---|---|
| Clone | K94011_7A4 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30208735
|
Invitrogen™
MA556089 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30213405
|
Invitrogen™
MA555709 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: KRKREQPDPS TKKQHTIWPS PHQGNSPDLE VYNVIRKQSE ADLAETRPDL KNISFRVCSG EATPDDMSCD YDNMAVNPSE SGFVTLVSVE SGFVTNDIYE FSPDQMGRSK
Layilin Monoclonal antibody specifically detects Layilin in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| Layilin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| E030012M19Rik; FLJ30977; FLJ31092; Gm511; layilin; LAYN; LYAN; RGD1564444; UNQ208/PRO234 | |
| Recombinant human LAYN (immunogen range is 261-370). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| LAYN |
| K94011_7A4 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6UX15 | |
| 143903 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title