missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Layilin Antibody (OTI4D3), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72403AF350
This item is not returnable.
View return policy
Description
Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for Western Blot.
Specifications
| Layilin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| LAYN | |
| Full length recombinant protein of human Layilin (NP_849156) produced in HEK293T cells. | |
| Signal Transduction | |
| 143903 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI4D3 | |
| Western Blot | |
| FLJ30977, FLJ31092, layilin | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction