missing translation for 'onlineSavingsMsg'
Learn More

Lawesson’s Reagent, TRC

Product Code. 30472148
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30472148 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30472148

Brand: Toronto Research Chemicals TRCL17878050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lawesson's Reagent
CAS 19172-47-5
Molecular Formula C14 H14 O2 P2 S4
Quantity 50 g
Synonym 2,4-Bis(4-methoxyphenyl)-1,3,2,4-dithiadiphosphetane 2,4-Disulfide, 2,4-Bis(4-methoxyphenyl)-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-Bis(4-methoxyphenyl)-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-Bis(4-methoxyphenyl)-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-Bis(p-methoxyphenyl)-1,3,2,4-dithiadiphosphetane 2,4-Disulfide, 2,4-Di(p-methoxyphenyl)-1,3-dithiadiphosphetane Disulfide, NSC 150550, Reagents, Lawesson's, p-Anisyldithiophosphonic Anhydride
InChI Formula InChI=1S/C14H14O2P2S4/c1-15-11-3-7-13(8-4-11)17(19)21-18(20,22-17)14-9-5-12(16-2)6-10-14/h3-10H,1-2H3
SMILES COc1ccc(cc1)P2(=S)SP(=S)(S2)c3ccc(OC)cc3
IUPAC Name 2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2λ^{5},4λ^{5}-dithiadiphosphetane
Molecular Weight (g/mol) 404.467
Formula Weight 403.935
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.