missing translation for 'onlineSavingsMsg'
Learn More

Lavendustin A, MedChemExpress

Product Code. 16433626
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
25 mg
missing translation for 'unitSize'
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16433626

missing translation for 'mfr': Medchem Express HY18963

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lavendustin A (RG-14355), isolated from Streptomyces Griseolavendus, is a potent, specific and ATP-competitive inhibitor of tyrosine kinase, with an IC50 of 11 ng/mL for EGFR-associated tyrosine kinase. It suppresses VEGF-induced angiogenesis and blocks the induction of LTPGABA-A.

Lavendustin A (RG-14355), isolated from Streptomyces Griseolavendus, is a potent, specific and ATP-competitive inhibitor of tyrosine kinase, with an IC50 of 11 ng/mL for EGFR-associated tyrosine kinase. It suppresses VEGF-induced angiogenesis and blocks the induction of LTPGABA-A.

Specifications

Chemical Name or Material Lavendustin A
CAS 125697-92-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H19NO6
Quantity 25 mg
Synonym RG-14355
Solubility Information DMSO : ≥ 250 mg/mL (655.51 mM)
SMILES O=C(O)C1=CC(N(CC2=CC(O)=CC=C2O)CC3=CC=CC=C3O)=CC=C1O
Molecular Weight (g/mol) 381.38
Formula Weight 381.38
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.