missing translation for 'onlineSavingsMsg'
Learn More

Lavendustin A, MedChemExpress

Product Code. 16487658
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487658 10 mg 10mg
16450128 5 mg 5mg
16433626 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16487658 Supplier Medchem Express Supplier No. HY18963
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lavendustin A (RG-14355), isolated from Streptomyces Griseolavendus, is a potent, specific and ATP-competitive inhibitor of tyrosine kinase, with an IC50 of 11 ng/mL for EGFR-associated tyrosine kinase. It suppresses VEGF-induced angiogenesis and blocks the induction of LTPGABA-A.

Lavendustin A (RG-14355), isolated from Streptomyces Griseolavendus, is a potent, specific and ATP-competitive inhibitor of tyrosine kinase, with an IC50 of 11 ng/mL for EGFR-associated tyrosine kinase. It suppresses VEGF-induced angiogenesis and blocks the induction of LTPGABA-A.

Specifications

Chemical Name or Material Lavendustin A
CAS 125697-92-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H19NO6
Quantity 10 mg
Synonym RG-14355
Solubility Information DMSO : ≥ 250 mg/mL (655.51 mM)
SMILES O=C(O)C1=CC(N(CC2=CC(O)=CC=C2O)CC3=CC=CC=C3O)=CC=C1O
Molecular Weight (g/mol) 381.38
Formula Weight 381.38
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.