missing translation for 'onlineSavingsMsg'
Learn More

Lauryl Methacrylate, TRC

Product Code. 30475585
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475585 1 g 1g
30466361 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30475585 Supplier Toronto Research Chemicals Supplier No. TRCL1787651G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-90-5
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
Synonym Ageflex FM 246, Blemmer LMA, Dodecyl 2-methyl-2-propenoate, Dodecyl methacrylate, Exceparl L-MA, GE 410, GE 410 (methacrylate), LAMA, LMA, Lauryl methacrylate, Light Ester L, MA 13, MA 13 (methacrylate), NSC 5188, Rocryl 320, SR 313, SR 313E, Sipomer LMA, n-Dodecyl methacrylate
IUPAC Name dodecyl 2-methylprop-2-enoate
SMILES CCCCCCCCCCCCOC(=O)C(=C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H30O2/c1-4-5-6-7-8-9-10-11-12-13-14-18-16(17)15(2)3/h2,4-14H2,1,3H3
Formula Weight 254.2246
Chemical Name or Material Lauryl Methacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.