missing translation for 'onlineSavingsMsg'
Learn More

Lauroyl chloride, 98%

Product Code. 10419270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10419270 250 g Glass bottle 250g
10308903 50 g Glass bottle 50g
10615612 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10419270 Supplier Thermo Scientific Acros Supplier No. 169032500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-16-3 | C12H23ClO | 218.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-16-3
Molecular Formula C12H23ClO
Molecular Weight (g/mol) 218.77
MDL Number MFCD00000740
InChI Key NQGIJDNPUZEBRU-UHFFFAOYSA-N
Synonym lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365
PubChem CID 8166
IUPAC Name dodecanoyl chloride
SMILES CCCCCCCCCCCC(Cl)=O

Specifications

Melting Point -17.0°C
Density 0.9200g/mL
Boiling Point 134.0°C to 137.0°C (11.0 mmHg)
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)10COCl
Refractive Index 1.4440 to 1.4460
Quantity 250 g
Beilstein 02, 363
Specific Gravity 0.92
Solubility Information Solubility in water: reacts. Other solubilities: organic solutions
Formula Weight 218.77
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Lauroyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lauroyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.